APO455835651 Apollo Scientific Ltd

2,6-Dichloro-5-fluoronicotinoyl chloride

  • MDL number MFCD01863240
  • Cas Number : 96568-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,6-Dichloro-5-fluoropyridine-3-carbonyl chloride, 3-(Chlorocarbonyl)-2,6-dichloro-5-fluoropyridine
Density 1.64
Flash Point 110
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 238-239
Product Link http://www.apolloscientific.co.uk/display_item.php?id=7634

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=7634
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart