APO455835653 Apollo Scientific Ltd

5-[4-(Trifluoromethyl)phenyl]-2H-tetrazole

  • MDL number MFCD00052525
  • Cas Number : 2251-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(2H-Tetrazol-5-yl)benzotrifluoride
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=7641

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=7641
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart