APO455835658 Apollo Scientific Ltd

3-Fluoro-4-nitropyridine

  • MDL number MFCD04112517
  • Cas Number : 13505-01-6
  • Molecular Weight : 142.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 93
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 62-64/5mm
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-nitropyridine

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-nitropyridine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart