APO455835659 Apollo Scientific Ltd

4-Chloro-2-(trifluoromethoxy)aniline

  • MDL number MFCD00153102
  • Cas Number : 175205-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-5-chloro-alpha,alpha,alpha-trifluoroanisole, 2-Amino-5-chlorophenyl trifluoromethyl ether
Purity 95
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 82/15mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=7731

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=7731
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart