APO455835662 Apollo Scientific Ltd

2-(2-Hydroxyethoxy)-4-(trifluoromethyl)pyridine

  • MDL number MFCD00126446
  • Cas Number : 257295-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-{[4-(Trifluoromethyl)pyridin-2-yl]oxy}ethan-1-ol, 2-(2-Hydroxyethoxy)-alpha,alpha,alpha-trifluoro-4-picoline
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=7744

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=7744
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart