APO455835664 Apollo Scientific Ltd

Ethyl 3-hydroxy-4,4,4-trifluorobutanoate

  • MDL number MFCD00040842
  • Cas Number : 372-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 23
Synonym 3-Hydroxy-4,4,4-trifluorobutyric acid ethyl ester
Purity 97
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 90-91/50mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-hydroxy-444-trifluorobutanoate-97

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-hydroxy-444-trifluorobutanoate-97
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart