APO455835666 Apollo Scientific Ltd

Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate

  • MDL number MFCD00219847
  • Cas Number : 175203-85-7
  • Molecular Weight : 301.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 297-305(dec.)
Synonym Ethyl 4-hydroxy-6-(trifluoromethoxy)-1-azanaphthalene-3-carboxylate, 3-(Ethoxycarbonyl)-4-hydroxy-6-(trifluoromethoxy)quinoline
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-hydroxy-6-trifluoromethoxyquinoline-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-hydroxy-6-trifluoromethoxyquinoline-3-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart