APO455835667 Apollo Scientific Ltd

4-Hydroxy-3-(trifluoromethyl)benzaldehyde

  • MDL number MFCD01091006
  • Cas Number : 220227-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-165
Synonym 4-Formyl-2-(trifluoromethyl)phenol, 5-Formyl-2-hydroxybenzotrifluoride
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3717

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3717
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart