APO455835670 Apollo Scientific Ltd

2-Amino-3-bromo-5-fluorobenzoic acid

  • MDL number MFCD00662509
  • Cas Number : 259269-84-6
  • Molecular Weight : 234.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-5-fluoroanthranilic acid, 2-Bromo-6-carboxy-4-fluoroaniline
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-3-bromo-5-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-3-bromo-5-fluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart