APO455835674 Apollo Scientific Ltd

5-Amino-2,2-difluoro-1,3-benzodioxole

  • MDL number MFCD00190144
  • Cas Number : 1544-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2-Difluoro-1,3-benzodioxol-5-amine, 3,4-(Difluoromethylenedioxy)aniline
Purity 98
Flash Point 85
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 95-97/12mm
Product Link https://store.apolloscientific.co.uk/product/5-amino-22-difluoro-13-benzodioxole-98

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-22-difluoro-13-benzodioxole-98
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart