APO455835678 Apollo Scientific Ltd

4-Fluoro-2-methyl-6-nitroaniline

  • MDL number MFCD09152778
  • Cas Number : 147285-87-8
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-100
Synonym 2-Amino-5-fluoro-3-nitrotoluene, 2-Amino-5-fluoro-3-methylnitrobenzene, 4-Fluoro-6-nitro-o-toluidine
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-2-methyl-6-nitroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-2-methyl-6-nitroaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart