APO455835692 Apollo Scientific Ltd

5-(Chlorosulphonyl)-2-fluorobenzoic acid

  • MDL number MFCD03030335
  • Cas Number : 37098-75-2
  • Molecular Weight : 238.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-113
Synonym 3-Carboxy-4-fluorobenzenesulphonyl chloride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chlorosulphonyl-2-fluorobenzoic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chlorosulphonyl-2-fluorobenzoic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart