APO455835694 Apollo Scientific Ltd

2-(Bromodifluoromethyl)-1,3-benzoxazole

  • MDL number MFCD11035866
  • Cas Number : 186828-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 62/1.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43523

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43523
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart