APO455835699 Apollo Scientific Ltd

2,2,2,2'-Tetrafluoroacetophenone

  • MDL number MFCD00040099
  • Cas Number : 124004-75-7
  • Molecular Weight : 192.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Fluorophenyl)-2,2,2-trifluoroethan-1-one
Purity 95
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 60-62/22mm
Product Link https://store.apolloscientific.co.uk/product/2222-tetrafluoroacetophenone

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2222-tetrafluoroacetophenone
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart