APO455835702 Apollo Scientific Ltd

2-Amino-6-iodo-4-[(trifluoromethyl)thio]phenol

  • MDL number MFCD25976388
  • Cas Number : 1448858-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-6-iodo-4-[(trifluoromethyl)sulphanyl]phenol, 2-Hydroxy-3-iodo-5-[(trifluoromethyl)thio]aniline
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=56517

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=56517
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart