APO455835703 Apollo Scientific Ltd

2-Bromo-5-fluorophenacyl bromide

  • MDL number MFCD23711741
  • Cas Number : 1427446-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-(2-bromo-5-fluorophenyl)ethan-1-one, 2,2'-Dibromo-5'-fluoroacetophenone
Purity 94
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=57170

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=57170
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart