APO455835705 Apollo Scientific Ltd

2-Fluoro-3-iodobenzoic acid

  • MDL number MFCD07774248
  • Cas Number : 447464-03-1
  • Molecular Weight : 266.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-iodobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-iodobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart