APO455835709 Apollo Scientific Ltd

3-Fluoro-2-iodobenzyl bromide

  • MDL number MFCD18396958
  • Cas Number : 1261675-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-3-fluoro-2-iodobenzene, alpha-Bromo-3-fluoro-2-iodotoluene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55739

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55739
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart