APO455835715 Apollo Scientific Ltd

4-(Difluoromethyl)benzaldehyde

  • MDL number MFCD18072541
  • Cas Number : 55805-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Formylbenzal fluoride, alpha,alpha-Difluoro-p-tolualdehyde
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59903

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=59903
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart