APO455835716 Apollo Scientific Ltd

2-Nitro-6-(trifluoromethyl)benzonitrile

  • MDL number MFCD16606330
  • Cas Number : 1227581-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Cyano-3-nitrobenzotrifluoride, 2-Cyano-3-(trifluoromethyl)nitrobenzene, 6-Nitro-alpha,alpha,alpha-trifluoro-o-tolunitrile
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60623

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=60623
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart