APO455835720 Apollo Scientific Ltd

4-Fluoro-2-formylbenzeneboronic acid

  • MDL number MFCD10697426
  • Cas Number : 825644-26-6
  • Molecular Weight : 167.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-125
Synonym 2-Borono-5-fluorobenzaldehyde
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-2-formylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-2-formylbenzeneboronic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart