APO455835722 Apollo Scientific Ltd

3-Fluoro-5-formylbenzeneboronic acid

  • MDL number MFCD11856017
  • Cas Number : 328956-60-1
  • Molecular Weight : 167.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Borono-5-fluorobenzaldehyde
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-formylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-formylbenzeneboronic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart