APO455835732 Apollo Scientific Ltd

5-Bromo-3-fluoro-2-nitrophenol

  • MDL number MFCD28348375
  • Cas Number : 1807256-50-3
  • Molecular Weight : 236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-3-fluoro-2-nitrophenol
Inchi Code 1S/C6H3BrFNO3/c7-3-1-4(8)6(9(11)12)5(10)2-3/h1-2,10H
Inchi Key CLCBYQYRMKRPAK-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-3-fluoro-2-nitrophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-3-fluoro-2-nitrophenol
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart