APO455835733 Apollo Scientific Ltd

Ethyl 5-(trifluoromethoxy)-1H-indole-2-carboxylate

  • MDL number MFCD00219837
  • Cas Number : 175203-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Ethoxycarbonyl)-5-(trifluoromethoxy)-1H-indole
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1654

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1654
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart