APO455835735 Apollo Scientific Ltd

Ethyl 2-(trifluoromethyl)phenylacetate

  • MDL number MFCD00236677
  • Cas Number : 81580-50-9
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(2-Ethoxy-2-oxoethyl)benzotrifluoride
Purity 97
Flash Point 66
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-trifluoromethylphenylacetate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-trifluoromethylphenylacetate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart