APO455835743 Apollo Scientific Ltd

6-(Trifluoromethyl)pyridine-3-boronic acid

  • MDL number MFCD07375381
  • Cas Number : 868662-36-6
  • Molecular Weight : 190.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Trifluoromethyl)pyridine-5-boronic acid, 5-Borono-2-(trifluoromethyl)pyridine
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-trifluoromethylpyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-trifluoromethylpyridine-3-boronic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart