APO455835745 Apollo Scientific Ltd

3-(Trifluoromethoxy)chlorobenzene

  • MDL number MFCD00276971
  • Cas Number : 772-49-6
  • Molecular Weight : 196.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.37
Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 140
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethoxychlorobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethoxychlorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart