APO455835750 Apollo Scientific Ltd

2,6-Difluoro-3-hydroxybenzeneboronic acid

  • MDL number MFCD09878345
  • Cas Number : 957065-86-0
  • Molecular Weight : 173.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-3-hydroxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-3-hydroxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart