APO455835752 Apollo Scientific Ltd

2,6-Difluoro-4-hydroxybenzeneboronic acid

  • MDL number MFCD09878344
  • Cas Number : 957065-87-1
  • Molecular Weight : 173.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157
Synonym 4-Borono-3,5-difluorophenol
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-4-hydroxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-4-hydroxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart