APO455835753 Apollo Scientific Ltd

2-Chloro-5-(trifluoromethyl)benzyl bromide

  • MDL number MFCD00236216
  • Cas Number : 237761-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Bromomethyl)-1-chloro-4-(trifluoromethyl)benzene, 3-(Bromomethyl)-4-chlorobenzotrifluoride
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 80/3mm
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-trifluoromethylbenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-trifluoromethylbenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart