APO455835755 Apollo Scientific Ltd

3'-Fluoro-4'-(trifluoromethyl)propiophenone

  • MDL number MFCD00236306
  • Cas Number : 237761-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-38
Synonym 1-[3-Fluoro-4-(trifluoromethyl)phenyl]propan-1-one, 2-Fluoro-4-propanoylbenzotrifluoride
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2241

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2241
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart