APO455835757 Apollo Scientific Ltd

3-Fluoro-2-nitropyridine

  • MDL number MFCD04114127
  • Cas Number : 54231-35-5
  • Molecular Weight : 142.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 30-35
Iupac Name 3-fluoro-2-nitropyridine
Inchi Code 1S/C5H3FN2O2/c6-4-2-1-3-7-5(4)8(9)10/h1-3H
Inchi Key IJVFHCSUEBAAOZ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 30-35
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-2-nitropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-2-nitropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart