APO455835777 Apollo Scientific Ltd

Diethyl 2,2-difluoromalonate

  • MDL number MFCD00221617
  • Cas Number : 680-65-9
  • Molecular Weight : 196.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Diethyl 2,2-difluoropropane-1,3-dioate
Purity 98
Density 1.17
Flash Point 62
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 94-95/23mm
Product Link https://store.apolloscientific.co.uk/product/diethyl-22-difluoromalonate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/diethyl-22-difluoromalonate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart