APO455835780 Apollo Scientific Ltd

2-Amino-3-bromo-5-(trifluoromethyl)pyridine

  • MDL number MFCD07375382
  • Cas Number : 79456-30-7
  • Molecular Weight : 241.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-101
Synonym 3-Bromo-5-(trifluoromethyl)pyridin-2-amine, 6-Amino-5-bromo-alpha,alpha,alpha-trifluoro-3-picoline
Purity 98
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-3-bromo-5-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-3-bromo-5-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart