APO455835785 Apollo Scientific Ltd

Methyl 4-fluoro-3-methylbenzoate

  • MDL number MFCD06203783
  • Cas Number : 180636-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 4-fluoro-m-toluate, 2-Fluoro-5-(methoxycarbonyl)toluene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-4-fluoro-3-methylbenzoate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-fluoro-3-methylbenzoate
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart