APO455835790 Apollo Scientific Ltd

4-Fluoro-2-iodoaniline

  • MDL number MFCD00042584
  • Cas Number : 61272-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.99
Flash Point >110
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 74-76
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5813

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5813
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart