APO455835801 Apollo Scientific Ltd

2,3-Difluoro-4-(trifluoromethyl)benzyl bromide

  • MDL number MFCD00236262
  • Cas Number : 213203-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Bromomethyl)-2,3-difluorobenzotrifluoride, 1-(Bromomethyl)-2,3-difluoro-4-(trifluoromethyl)benzene
Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1494

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1494
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart