APO455835802 Apollo Scientific Ltd

4-Fluorobenzamide

  • MDL number MFCD00007992
  • Cas Number : 824-75-9
  • Molecular Weight : 139.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 154-156
Iupac Name 4-fluorobenzamide
Inchi Code 1S/C7H6FNO/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H2,9,10)
Inchi Key VNDHYTGVCGVETQ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 154-156
Product Link https://store.apolloscientific.co.uk/product/4-fluorobenzamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluorobenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart