APO455835805 Apollo Scientific Ltd

4-Methoxy-2-(trifluoromethyl)benzaldehyde

  • MDL number MFCD01091011
  • Cas Number : 106312-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-41
Synonym 2-Formyl-5-methoxybenzotrifluoride, 4-Formyl-3-(trifluoromethyl)anisole
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5815

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5815
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart