APO455835817 Apollo Scientific Ltd

4-Methoxy-2-(trifluoromethyl)benzoic acid

  • MDL number MFCD01091013
  • Cas Number : 127817-85-0
  • Molecular Weight : 220.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-150
Synonym 2-(Trifluoromethyl)-p-anisic acid, 2-Carboxy-5-methoxybenzotrifluoride, 4-Carboxy-3-(trifluoromethyl)anisole
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methoxy-2-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methoxy-2-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart