APO455835826 Apollo Scientific Ltd

3-Fluoro-4-(methylcarbamoyl)benzeneboronic acid

  • MDL number MFCD08436026
  • Cas Number : 849833-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-175
Synonym 4-Borono-2-fluoro-N-methylbenzamide
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-methylcarbamoylbenzeneboronic-acid-98

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-methylcarbamoylbenzeneboronic-acid-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart