APO455835827 Apollo Scientific Ltd

3-Fluoropyridine-4-boronic acid

  • MDL number MFCD03788558
  • Cas Number : 458532-97-3
  • Molecular Weight : 140.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-239
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoropyridine-4-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoropyridine-4-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart