APO455835828 Apollo Scientific Ltd

2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid

  • MDL number MFCD08436034
  • Cas Number : 603122-84-5
  • Molecular Weight : 197.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-175
Synonym Methyl 4-borono-3-fluorobenzoate
Purity 96
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-methoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-methoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart