APO455835838 Apollo Scientific Ltd

4-Fluoro-3-hydroxybenzeneboronic acid

  • MDL number MFCD08689483
  • Cas Number : 913835-74-2
  • Molecular Weight : 155.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 264-266
Synonym 5-Borono-2-fluorophenol
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-hydroxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-hydroxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart