APO455835847 Apollo Scientific Ltd

Methyl 4-bromo-3-fluorobenzoate

  • MDL number MFCD08436044
  • Cas Number : 849758-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-66
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-4-bromo-3-fluorobenzoate-98

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-bromo-3-fluorobenzoate-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart