APO455835863 Apollo Scientific Ltd

4,4,4-Trifluorobutylamine hydrochloride

  • MDL number MFCD09258951
  • Cas Number : 84153-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-186
Synonym 4,4,4-Trifluorobutan-1-amine hydrochloride
Purity 95
Flash Point >110
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11236

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=11236
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart