APO455835870 Apollo Scientific Ltd

Methyl 3-bromo-2-fluorobenzoate

  • MDL number MFCD09261255
  • Cas Number : 206551-41-9
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-37
Physical Form Solid-Fused
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-bromo-2-fluorobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-bromo-2-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart