APO455835871 Apollo Scientific Ltd

4-Amino-2-fluorobenzotrifluoride

  • MDL number MFCD00190120
  • Cas Number : 69411-68-3
  • Molecular Weight : 179.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-58
Synonym 3-Fluoro-4-(trifluoromethyl)aniline, alpha,alpha,alpha,3-Tetrafluoro-p-toluidine
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-2-fluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-2-fluorobenzotrifluoride
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart