APO455835874 Apollo Scientific Ltd

4-Bromo-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD04969230
  • Cas Number : 19968-17-3
  • Molecular Weight : 214.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-114
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart