APO455835879 Apollo Scientific Ltd

2,3-Dichloro-4-fluoronitrobenzene

  • MDL number MFCD09763598
  • Cas Number : 36556-51-1
  • Molecular Weight : 209.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3-Dichloro-1-fluoro-4-nitrobenzene
Density 1.60
Flash Point >110
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-dichloro-4-fluoronitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-dichloro-4-fluoronitrobenzene
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart